Lithiumbis(trimethylsilyl)amide 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lithiumbis(triméthylsilyl)amide, HMDS-Li, N-Lithiohexamethyldisilazane, HMDS lithium salt

AC33814-0100 AC33814-0500 AC33814-2500
TS33814-0100EA 38 USD
TS33814-0100 TS33814-0500 TS33814-2500
Lithiumbis(trimethylsilyl)amide 95%
Lithiumbis(trimethylsilyl)amide
Formula: C₆H₁₈LiNSi₂
MW: 167.33 g/mol
Boiling Pt: 114…116 °C
Density: 0.891 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00008261
CAS Number: 4039-32-1
EINECS: 223-725-6
UN: 2925
ADR: 4.1 II

Order Now

Specification Test Results

Appearance Yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration with HClO4 ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR