Potassium thiocyanate 98%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC37743-0025
AC37743-1000
AC37743-5000
TS37743-0025EA
169
USD
TS37743-0025
TS37743-1000
TS37743-5000
Potassium thiocyanate 98%, pure
Potassium thiocyanate
Formula:
KSCN MW: 97.18 g/mol Melting Pt: 170…179 °C Density: 1.886 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011413 CAS Number: 333-20-0 EINECS: 206-370-1 Merck Index: 12,07866 |
Specification Test Results
Appearance | Colorless to white Crystalline powder or crystals |
Titration Argentometric | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...