Potassium disulfite 97%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

AC41827-0025 AC41827-0250 AC41827-5000
TS41827-0025EA 132 USD
TS41827-0025 TS41827-0250 TS41827-5000
Potassium disulfite 97%, Extra Pure
Potassium disulphite
Formula: K₂O₅S₂
Melting Pt: 150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00167605
CAS Number: 16731-55-8
EINECS: 240-795-3
Merck Index: 13,07729

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Titration Iodimetric ≥96.0 %
Arsenic (As) ≤1 ppm
Iron (Fe) ≤5 ppm
Selenium (Se) ≤1 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR