Diammonium thiosulfate 96%, pure
Supplier: Thermo Scientific Chemicals
AC39166-0025
AC39166-0100
AC39166-5000
TS39166-0025EA
166
USD
TS39166-0025
TS39166-0100
TS39166-5000
Diammonium thiosulfate 96%, pure
Diammonium thiosulphate
Formula:
H₈N₂O₃S₂ Melting Pt: 150 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00015965 CAS Number: 7783-18-8 |
Specification Test Results
Appearance | White Crystalline powder or crystals and/or chunks |
Titration Iodimetric | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...