Molybdenum(IV) sulphide 98.5%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Molybdenum disulphide

AC21578-0025 AC21578-1000 AC21578-5000
TS21578-0025EA 717 USD
TS21578-0025 TS21578-1000 TS21578-5000
Molybdenum(IV) sulphide 98.5%
Molybdenum(IV) sulphide
Formula: MoS₂
Melting Pt: 2375 °C
Storage Temperature: Ambient
MDL Number: MFCD00003470
CAS Number: 1317-33-5
EINECS: 215-263-9
Merck Index: 12,06318

Order Now

Specification Test Results

Appearance Gray to black Powder
Assay ≥98.0 % (MoS2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR