Ammonium dihydrogen phosphate 99.9% (trace metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

AC19370-0025 AC19370-1000 AC19370-5000
TS19370-0025EA 258 USD
TS19370-0025 TS19370-1000 TS19370-5000
Ammonium dihydrogen phosphate 99.9% (trace metals basis)
Ammonium dihydrogen phosphate
Formula: NH₄H₂PO₄
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5
Merck Index: 13,00547

Order Now

Specification Test Results

Appearance White Crystalline powder
Total trace metal impurities ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR