Diammonium hydrogen orthophosphate 99% for biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

AC44714-0010 AC44714-2500
TS44714-0010EA 142 USD
TS44714-0010 TS44714-2500
Diammonium hydrogen orthophosphate 99% for biochemistry
Diammonium hydrogen orthophosphate
Formula: (NH₄)₂HPO₄
MW: 132.06 g/mol
Melting Pt: 185
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS Number: 7783-28-0
Merck Index: 13,00546

Order Now

Specification Test Results

Appearance White Crystals
Solubility (1 M in water) Clear colorless
UV at 280 nm A: ≤0.1
pH 7.5 to 9.0 (1 M in water)
Titration with HCl ≥98.5 %
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR