Calcium hydrogen orthophosphate, anhydrous 98+% for analysis

Supplier: Thermo Scientific Chemicals

Synonyms: Calcium phosphate dibasic, Calcium hydrogen phosphate, Calcium perphosphate, ammonified, Anhydrous Dibasic Calcium Phosphate, Calcium phosphate, dibasic

AC44643-0500 AC44643-2500
TS44643-0500EA 40 USD
TS44643-0500 TS44643-2500
Calcium hydrogen orthophosphate, anhydrous 98+% for analysis
Calcium hydrogen orthophosphate
Formula: CaHPO₄
MW: 136.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010909
CAS Number: 7757-93-9

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric 98.0 to 103.0 %
Chloride (Cl) ≤0.033 %
Fluoride (F) ≤0.005 %
Carbonate (CO3) Passes test
Sulfate (SO4) ≤0.5 %
Arsenic (As) ≤0.0003 %
Barium (Ba) Passes test
Iron (Fe) ≤0.04 %
Insoluble matter ≤0.2 % (in HCl)
Loss on ignition 6.6 % to 8.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR