Calcium hydrogen orthophosphate, anhydrous 97%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Calcium phosphate dibasic, Calcium hydrogen phosphate, Calcium perphosphate, ammonified, Anhydrous Dibasic Calcium Phosphate, Calcium phosphate, dibasic

AC21571-0010 AC21571-0050 AC21571-0250
TS21571-0010EA 95 USD
TS21571-0010 TS21571-0050 TS21571-0250
Calcium hydrogen orthophosphate, anhydrous 97%, pure
Calcium hydrogen orthophosphate
Formula: CaHPO₄
MW: 136.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010909
CAS Number: 7757-93-9
EINECS: 231-826-1
Merck Index: 13,01697

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR