Tripotassium orthophosphate 90+ %, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium phosphate tribasic, Potassium phosphate

AC46733-0010 AC46733-0050 AC46733-0250
TS46733-0010EA 55 USD
TS46733-0010 TS46733-0050 TS46733-0250
Tripotassium orthophosphate 90+ %, pure
Tripotassium orthophosphate
Formula: K₃O₄P
Melting Pt: 1340 °C
Storage Temperature: Ambient
MDL Number: MFCD00036295
CAS Number: 7778-53-2
EINECS: 231-907-1
Merck Index: 12,07830

Order Now

Specification Test Results

Appearance (Color) White
Appearance (Form) Powder or granules and/or chunks
Titration excess HCl ≥90.0 %
Water =<10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR