Tripotassium orthophosphate, anhydrous 97%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Potassium phosphate tribasic, Potassium phosphate
AC38768-0010
AC38768-0025
AC38768-0100
AC38768-0250
AC38768-5000
TS38768-0010EA
93
USD
TS38768-0010
TS38768-0025
TS38768-0100
TS38768-0250
TS38768-5000
Tripotassium orthophosphate, anhydrous 97%, pure
Tripotassium orthophosphate
Formula:
K₃O₄P MW: 212.27 g/mol Melting Pt: 1340 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036295 CAS Number: 7778-53-2 EINECS: 231-907-1 Merck Index: 12,07830 |
Specification Test Results
Appearance | White Powder or granules and/or chunks |
Titration excess HCl | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...