Cobalt(II) hydroxide 95%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobalt dihydroxide, Cobaltous hydroxide, Cobalt(II) hydroxide

AC45389-0010 AC45389-2500
TS45389-0010EA 346 USD
TS45389-0010 TS45389-2500
Cobalt(II) hydroxide 95%, pure
Cobalt(II) hydroxide
Formula: H₂CoO₂
MW: 92.95
Density: 3.597
Storage Temperature: Ambient
MDL Number: MFCD00016015
CAS Number: 21041-93-0
UN: 3077
ADR: 9 III
Merck Index: 13,02467

Order Now

Specification Test Results

Appearance Pink to red Powder
Titration excess HCl ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR