Triethoxysilane 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC16481-0100
AC16481-0500
AC16481-2500
TS16481-0100EA
59
USD
TS16481-0100
TS16481-0500
TS16481-2500
Triethoxysilane 95%
Triethoxysilane
Formula:
C₆H₁₆O₃Si MW: 164.28 g/mol Boiling Pt: 135 Melting Pt: -170 Density: 0.89 g/cm³ Flash Pt: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009061 CAS Number: 998-30-1 UN: 3489 ADR: 6.1 I |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3770 to 1.3790 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...