Dibutyltin oxide 98%

Supplier: Thermo Scientific Chemicals
Danger

AC17936-1000 AC17936-5000
TS17936-1000EA 38 USD
TS17936-1000 TS17936-5000
Dibutyltin oxide 98%
Dibutyltin oxide
Formula: C₈H₁₈OSn
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00001992
CAS Number: 818-08-6
EINECS: 212-449-1
UN: 3146
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White Powder or granular powder
Infrared spectrum Conforms
Chloride (Cl) ≤0.2 %
Tin (Sn) ≥47 %
Volatile matter ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR