Cerium(IV) oxide 99.9% (trace metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Ceric oxide, Cerium dioxide

AC19912-0025 AC19912-1000 AC19912-5000
TS19912-0025EA 659 USD
TS19912-0025 TS19912-1000 TS19912-5000
Cerium(IV) oxide 99.9% (trace metals basis)
Cerium(IV) oxide
Formula: CeO₂
MW: 172.11 g/mol
Boiling Pt: 3468
Melting Pt: 2000
Density: 7.13 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035

Order Now

Specification Test Results

Appearance White to yellow Powder (May give a pink to red-brown color due to traces lanthanum, neodymium, praseodymium, etc.)
Total conc. of imp.as REO ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR