Antimony(III) oxide 99+%

Supplier: Thermo Scientific Chemicals
Warning

AC21347-0010 AC21347-0051 AC21347-1000
TS21347-0010EA 157 USD
TS21347-0010 TS21347-0051 TS21347-1000
Antimony(III) oxide 99+%
Antimony(III) oxide
Formula: O₃Sb₂
Boiling Pt: 1550 °C (760 mmHg)
Melting Pt: 656 °C
Storage Temperature: Ambient
MDL Number: MFCD00011214
CAS Number: 1309-64-4
EINECS: 215-175-0
Merck Index: 13,00717

Order Now

Specification Test Results

Appearance White Powder
Assay ≥99.0 % (Sb2O3)
Particle size ≤0.01 % Residue on 45 µm sieve
Average particle size 0.8 to 1.0 µm
Arsenic (As) ≤500 ppm
Iron (Fe) ≤30 ppm
Lead (Pb) ≤400 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR