Ammonium cerium(IV) nitrate 99% for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC20144-0010
AC20144-0050
AC20144-1000
AC20144-2500
TS20144-0010EA
462
USD
TS20144-0010
TS20144-0050
TS20144-1000
TS20144-2500
Ammonium cerium(IV) nitrate 99% for analysis
Ammonium cerium(IV) nitrate
Formula:
(NH₄)₂Ce(NO₃)₆ Melting Pt: 107…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151121 CAS Number: 16774-21-3 EINECS: 240-827-6 UN: 3085 ADR: 5.1,II Merck Index: 13,00509 |
Specification Test Results
Appearance | Orange Crystalline powder or crystals |
Solubility | (10 % in water) Clear |
Moisture | ≤2 % |
Titration | ≥98.5 % (on dry substance) |
Calcium oxide (CaO) | ≤5 ppm |
Iron(III)oxide (Fe2O3) | ≤5 ppm |
Potassium oxide (K2O) | ≤3 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...