Bismuth(III) nitrate pentahydrate 99.999% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Bismuth trinitrate pentahydrate
AC31515-0050
AC31515-0250
AC31515-1000
TS31515-0050EA
35
USD
TS31515-0050
TS31515-0250
TS31515-1000
Bismuth(III) nitrate pentahydrate 99.999% (trace metals basis)
Bismuth(III) nitrate pentahydrate
Formula:
Bi(NO₃)₃·5H₂O MW: 485.07 g/mol Boiling Pt: 80 °C (760 mmHg) Melting Pt: 30 Storage Temperature: Ambient |
MDL Number:
MFCD00149157 CAS Number: 10035-06-0 EINECS: 233-791-8 UN: 1477 ADR: 5.1 II |
Specification Test Results
Appearance | White Crystals |
Total trace metal impurities | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...