N-(3-(Trimethoxysilyl)propyl)ethylenediamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(2-Aminoethylamino)propyltrimethoxysilane, N-(2-Aminoethyl)-3-aminopropyltrimethoxysilane, N-[3-(Trimethoxysilyl)propyl]ethylenediamine

AC21653-1000 AC21653-5000
TS21653-1000EA 71 USD
TS21653-1000 TS21653-5000
N-(3-(Trimethoxysilyl)propyl)ethylenediamine 97%
N-(3-(Trimethoxysilyl)propyl)ethylenediamine
Formula: H₂NCH₂CH₂NHCH₂CH₂CH₂Si(OCH₃)₃
MW: 222.36 g/mol
Boiling Pt: 261…263 °C (1013 hPa)
Melting Pt: ∼ 0 °C
Density: 1.03 g/cm³
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00008173
CAS Number: 1760-24-3
EINECS: 217-164-6

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4425 to 1.4445 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR