(3-Aminopropyl)triethoxysilane 99%, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Aminopropyltriethoxysilane, 3-(Triethoxysilyl)propylamine, Geniosil GF 93, Triethoxy(3-aminopropyl)silane
AC43094-1000
AC43094-8000
TS43094-1000EA
82
USD
TS43094-1000
TS43094-8000
(3-Aminopropyl)triethoxysilane 99%, AcroSeal™
(3-Aminopropyl)triethoxysilane
Formula:
H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃ MW: 221.37 g/mol Boiling Pt: 217 °C (760 mmHg) Melting Pt: –70 °C Density: 0.942 g/cm³ Flash Pt: 96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008207 CAS Number: 919-30-2 UN: 2735 ADR: 8,II |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4200 to 1.4220 (20°C, 589 nm) |
GC | ≥98.5 % |
Color scale | ≤25 APHA |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...