Molybdic acid for analysis
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC21362-1000
AC21362-5000
TS21362-1000EA
79
USD
TS21362-1000
TS21362-5000
Molybdic acid for analysis
Molybdic acid
Formula:
H₂MoO₄ Boiling Pt: ∼300 °C (1013 hPa) Melting Pt: 300 °C Density: 3.1 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00036287 CAS Number: 7782-91-4 EINECS: 231-970-5 Merck Index: 13,06262 |
Specification Test Results
Appearance | White to light yellow Powder |
Additional info | contains ammonium molybdate |
Titration with NaOH | ≥85 % (as MoO3) |
Chloride (Cl) | ≤100 ppm |
Phosphate (PO4) | ≤5 ppm |
Sulfate (SO4) | ≤2000 ppm |
Iron (Fe) | ≤50 ppm |
Heavy metals (as Pb) | ≤30 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...