Tetrabutylammonium iodide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetra-n-butylammonium iodide, TBAI

AC16127-0025 AC16127-0250 AC16127-1000 AC16127-5000
TS16127-0025EA 1035 USD
TS16127-0025 TS16127-0250 TS16127-1000 TS16127-5000
Tetrabutylammonium iodide 98%
Tetrabutylammonium iodide
Formula: C₁₆H₃₆IN
MW: 369.37 g/mol
Melting Pt: 141…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00011636
CAS Number: 311-28-4
EINECS: 206-220-5

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 141 to 150 °C
Titration Argentometric ≥97.5 %
Loss on drying ≤1 %
Residue after ignition ≤1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR