Cadmium(II) iodide 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cadmium iodide

AC21146-0010 AC21146-2500
TS21146-0010EA 674 USD
TS21146-0010 TS21146-2500
Cadmium(II) iodide 99%
Cadmium(II) iodide
Formula: CdI₂
MW: 366.22 g/mol
Boiling Pt: 787 °C (760 mmHg)
Melting Pt: 388 °C
Storage Temperature: Ambient
MDL Number: MFCD00010920
CAS Number: 7790-80-9
EINECS: 232-223-6
UN: 2570
ADR: 6.1,III
Merck Index: 12,01657

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals and/or flakes
Titration Complexometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR