Copper(I) iodide 98%

Supplier: Thermo Scientific Chemicals
Danger

AC19490-0010 AC19490-0050 AC19490-2500
TS19490-0010EA 264 USD
TS19490-0010 TS19490-0050 TS19490-2500
Copper(I) iodide 98%
Copper(I) iodide
Formula: CuI
Boiling Pt: 1290 °C (1013 hPa)
Melting Pt: 605 °C
Storage Temperature: Ambient
MDL Number: MFCD00010978
CAS Number: 7681-65-4
EINECS: 231-674-6
UN: 3077
ADR: 9,III
Merck Index: 13,02692

Order Now

Specification Test Results

Appearance Off-white to gray-brown Powder
Titration Complexometric ≥97.5 %
Sulfate (SO4) ≤5000 ppm
Iron (Fe) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR