Cesium iodide 99.9% for analysis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cesiumiodide

AC19205-0250 AC19205-1000
TS19205-0250EA 82 USD
TS19205-0250 TS19205-1000
Cesium iodide 99.9% for analysis
Cesium iodide
Formula: CsI
MW: 259.81 g/mol
Boiling Pt: 1280 °C (1013 hPa)
Melting Pt: 626 °C
MDL Number: MFCD00010962
CAS Number: 7789-17-5
EINECS: 232-145-2
UN: 3077
Merck Index: 12,02056

Order Now

Specification Test Results

Appearance White Powder
Purity ≥99.85 % (metals basis)
Aluminium (Al) ≤10 ppm
Bismuth (Bi) ≤5 ppm
Cadmium (Cd) ≤5 ppm
Calcium (Ca) ≤10 ppm
Chromium (Cr) ≤5 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤5 ppm
Lithium (Li) ≤10 ppm
Nickel (Ni) ≤5 ppm
Potassium (K) ≤50 ppm
Rubidium (Rb) ≤50 ppm
Silicon (Si) ≤50 ppm
Sodium (Na) ≤50 ppm
Zinc (Zn) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR