Cobalt(II) fluoride 99%
Supplier: Thermo Scientific Chemicals
AC31127-0050
AC31127-0250
TS31127-0050EA
33
USD
TS31127-0050
TS31127-0250
Cobalt(II) fluoride 99%
Cobalt(II) fluoride
Specification Test Results
Appearance | Pink Powder |
Assay | ≥98.5 % (ex F) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...