Aluminium fluoride trihydrate 97%, extra pure

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminum fluoride trihydrate

AC21682-0025 AC21682-5000
TS21682-0025EA 406 USD
TS21682-0025 TS21682-5000
Aluminium fluoride trihydrate 97%, extra pure
Aluminium fluoride trihydrate
Formula: AlF₃·3H₂O
MW: 138.02
Density: 1.91
Storage Temperature: Ambient
MDL Number: MFCD00149141
CAS Number: 15098-87-0
EINECS: 232-051-1
Merck Index: 12,00351

Order Now

Specification Test Results

Appearance White Powder
Assay ≥96.0 %
Sulfate (SO4) ≤200 ppm
Iron (Fe) ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR