Copper(II) fluoride, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cupric fluoride

AC20337-0250 AC20337-1000
TS20337-0250EA 150 USD
TS20337-0250 TS20337-1000
Copper(II) fluoride, anhydrous 98%
Copper(II) fluoride
Formula: CuF₂
MW: 101.54 g/mol
Boiling Pt: 950 °C (1013 hPa)
Melting Pt: 950 °C (dec.)
Density: 4.23 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010977
CAS Number: 7789-19-7
EINECS: 232-147-3
UN: 3260
ADR: 8,II
Merck Index: 12,02704

Order Now

Specification Test Results

Appearance Light gray to blue-gray Powder
Assay ≥97.5 %
Loss on drying ≤0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR