Cesium fluoride 99.9%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

AC31591-0050 AC31591-0250
TS31591-0050EA 45 USD
TS31591-0050 TS31591-0250
Cesium fluoride 99.9%, Extra Pure
Cesium fluoride
Formula: CsF
MW: 151.9 g/mol
Boiling Pt: 1251 °C (1013 hPa)
Melting Pt: 682 °C
Storage Temperature: Ambient
MDL Number: MFCD00010960
CAS Number: 13400-13-0
EINECS: 236-487-3
UN: 3288
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White Powder
Total trace metal impurities ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR