Ammonium dichromate 99%

Supplier: Thermo Scientific Chemicals
Danger

AC20881-0025 AC20881-1000 AC20881-5000
TS20881-0025EA 483 USD
TS20881-0025 TS20881-1000 TS20881-5000
Ammonium dichromate 99%
Ammonium dichromate
Formula: (NH₄)₂Cr₂O₇
MW: 252.06 g/mol
Melting Pt: 170 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010879
CAS Number: 7789-09-5
EINECS: 232-143-1
UN: 1439
ADR: 5.1,II
REACH: 01-2119661563-36-0000
Merck Index: 13,00516

Order Now

Specification Test Results

Appearance Orange Adhering crystals
Titration Iodometric ≥98.5 %
Loss on drying ≤3 % (105°C, 4 h)
Chloride (Cl) ≤500 ppm
Sulfate (SO4) ≤500 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR