Triammonium citrate 97+%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-1,2,3-propanetricarboxylic acid triammonium salt, Ammonium citrate tribasic, Citric acid triammonium salt, Triammonium 2-hydroxypropane-1,2,3-tricarboxylate

AC39141-0010 AC39141-0500 AC39141-2500
TS39141-0010EA 252 USD
TS39141-0010 TS39141-0500 TS39141-2500
Triammonium citrate 97+%, pure
Triammonium citrate
Formula: C₆H₁₇N₃O₇
MW: 243.22 g/mol
Melting Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00036406
CAS Number: 3458-72-8

Order Now

Specification Test Results

Appearance White to off-white Crystals
Infrared spectrum Conforms
Titration with HClO4 ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR