Diammonium hydrogen citrate 98+% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Citric acid diammonium salt, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Ammonium citrate dibasic

AC44768-0025 AC44768-1000 AC44768-5000
TS44768-0025EA 646 USD
TS44768-0025 TS44768-1000 TS44768-5000
Diammonium hydrogen citrate 98+% ACS
Diammonium hydrogen citrate
Formula: HOC(CO₂H)(CH₂CO₂NH₄)₂
MW: 226.19 g/mol
Density: 1.5 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00013068
CAS Number: 3012-65-5
Merck Index: 13,00513

Order Now

Specification Test Results

Appearance White Crystalline powder
Residue after ignition ≤0.01 %
Titration with NaOH 98.0 to 103.0 %
Chloride (Cl) ≤0.001 %
Oxalate (C2O4) Passes test (Limit about 0.05%)
Phosphate (PO4) ≤5 ppm
Iron (Fe) ≤0.001 %
Heavy metals (ICP-OES) ≤5 ppm
Sulfur compounds (as SO4) ≤0.005 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR