L(+)-Potassium hydrogen tartrate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Potassium hydrogen L-tartrate, Potassium acid tartrate, Potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Potassium Bitartrate, (R,R)-(+)-Tartaric acid potassium salt, L(+)-Tartaric acid potassium salt
AC22267-0010
AC22267-0051
AC22267-2500
TS22267-0010EA
117
USD
TS22267-0010
TS22267-0051
TS22267-2500
L(+)-Potassium hydrogen tartrate 99%
L(+)-Potassium hydrogen tartrate
Formula:
KOOC(CHOH)₂COOH MW: 188.18 g/mol Melting Pt: 250 Density: 1.95 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00065392 CAS Number: 868-14-4 EINECS: 212-769-1 Merck Index: 13,07697 |
|
Specification Test Results
Appearance | Colorless to white Powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (105°C) |
Specific optical rotation | +8.0° to +9.2° (20°C, 589 nm) (c=10, HCl 1M and H2O) (on dried substance) |
Titration with NaOH | 98.5 to 101.5 % (On dried substance) |
Chloride (Cl) | ≤500 ppm |
Sulfate (SO4) | ≤500 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...