di-Potassium oxalate monohydrate 99% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Oxalic acid potassium salt monohydrate, Potassium oxalate hydrate, Potassium oxalate monohydrate, Potassium ethanedioate monohydrate
AC42402-0025
AC42402-5000
TS42402-0025EA
295
USD
TS42402-0025
TS42402-5000
di-Potassium oxalate monohydrate 99% ACS
di-Potassium oxalate monohydrate
Formula:
(COOK)₂·H₂O MW: 184.23 g/mol Melting Pt: 356 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150033 CAS Number: 6487-48-5 EINECS: 209-506-8 Merck Index: 12,07820 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration with KMnO4 | 98.5 to 101.0 % |
Substance darkened by H2SO4 | Passes test |
Chloride (Cl) | ≤20 ppm |
Sulfate (SO4) | ≤100 ppm |
Ammonium (NH4) | ≤20 ppm |
Iron (Fe) | ≤10 ppm |
Sodium (Na) | ≤200 ppm |
Heavy metals (as Pb) | ≤20 ppm |
Insoluble matter | ≤0.01 % |
Neutrality | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...