Cadmium acetate dihydrate 98%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cadmium di(acetate) dihydrate, Acetic acid cadmium salt dihydrate

AC21231-0010 AC21231-0050 AC21231-2500
TS21231-0010EA 175 USD
TS21231-0010 TS21231-0050 TS21231-2500
Cadmium acetate dihydrate 98%, pure
Cadmium acetate dihydrate
Formula: (H₃CCOO)₂Cd·2H₂O
MW: 266.53 g/mol
Boiling Pt: 105…107 °C (4 mmHg)
Melting Pt: 256 °C
Storage Temperature: Ambient
MDL Number: MFCD00150020
CAS Number: 5743-04-4
UN: 2570
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White Crystals
Titration Complexometric ≥97.5 %
Loss on drying 12.5 % to 14.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR