Cadmium acetate dihydrate 98% for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cadmium di(acetate) dihydrate, Acetic acid cadmium salt dihydrate

AC31713-0025 AC31713-1000 AC31713-5000
TS31713-0025EA 892 USD
TS31713-0025 TS31713-1000 TS31713-5000
Cadmium acetate dihydrate 98% for analysis
Cadmium acetate dihydrate
Formula: (H₃CCOO)₂Cd·2H₂O
MW: 266.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150020
CAS Number: 5743-04-4
EINECS: 208-853-2
UN: 2570
ADR: 6.1,III
Merck Index: 13,01614

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Complexometric ≥97.5 %
Chloride (Cl) ≤10 ppm
Nitrate (NO3) ≤30 ppm
Sulfate (SO4) ≤50 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤50 ppm
Zinc (Zn) ≤50 ppm
Insoluble matter ≤0.005 % (in water)
Subst not pptd by H2S ≤0.3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR