Copper(II) acetate monohydrate ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate

AC42359-0025 AC42359-1000 AC42359-5000
TS42359-0025EA 449 USD
TS42359-0025 TS42359-1000 TS42359-5000
Copper(II) acetate monohydrate ACS
Copper(II) acetate monohydrate
Formula: (H₃CCOO)₂Cu·H₂O
MW: 199.65 g/mol
Boiling Pt: 240 °C (760 mmHg)
Melting Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00149570
CAS Number: 6046-93-1
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance Blue-green to dark green Solid
Infrared spectrum Conforms
Titration Iodometric 98.0 to 102.0 %
Chloride (Cl) ≤30 ppm
Sulfate (SO4) ≤100 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤20 ppm
Nickel (Ni) ≤100 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤500 ppm
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR