Sodium acetate trihydrate 99+% for biochemistry
Supplier: Thermo Scientific Chemicals
Synonyms:
Acetic acid sodium salt trihydrate
AC44679-0010
AC44679-2500
TS44679-0010EA
70
USD
TS44679-0010
TS44679-2500
Sodium acetate trihydrate 99+% for biochemistry
Sodium acetate trihydrate
Formula:
H₃CCOONa·3H₂O MW: 136.08 g/mol Melting Pt: 58 Storage Temperature: Ambient |
MDL Number:
MFCD00071557 CAS Number: 6131-90-4 Merck Index: 13,08642 |
|
Specification Test Results
Appearance | White Crystals |
Infrared spectrum | Conforms |
UV | at 280 nm A: ≤0.05 |
pH | 7.5 to 9.5 |
Titration with HClO4 | 99.0 to 101.0 % |
Chloride (Cl) | ≤0.001 % |
Phosphate (PO4) | ≤0.0005 % |
Sulfate (SO4) | ≤0.002 % |
Calcium (Ca) | ≤0.0005 % |
Iron (Fe) | ≤0.0005 % |
Magnesium (Mg) | ≤0.0005 % |
Potassium (K) | ≤0.005 % |
Zinc (Zn) | ≤0.0005 % |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % (in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...