Sodium butyrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Butyric acid sodium salt, Sodium Butyrate

AC26319-0050 AC26319-0250 AC26319-1000 AC26319-5000
TS26319-0050EA 26 USD
TS26319-0050 TS26319-0250 TS26319-1000 TS26319-5000
Sodium butyrate 98%
Sodium butyrate
Formula: C₄H₇NaO₂
MW: 110.09 g/mol
Melting Pt: 250…253 °C
Storage Temperature: Ambient
MDL Number: MFCD00002816
CAS Number: 156-54-7
EINECS: 205-857-6

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR