Benzoyl bromide 97%

Supplier: Thermo Scientific Chemicals
Danger

AC16095-0050 AC16095-0250 AC16095-1000
TS16095-0050EA 67 USD
TS16095-0050 TS16095-0250 TS16095-1000
Benzoyl bromide 97%
Benzoyl bromide
Formula: C₇H₅BrO
MW: 185.02 g/mol
Boiling Pt: 218
Melting Pt: 0
Density: 1.57 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00000112
CAS Number: 618-32-6
EINECS: 210-544-2
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.5880 to 1.5920 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR