Ammonium hydrogen carbonate 98%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ammonium bicarbonate
AC37093-0010
AC37093-0050
AC37093-0250
TS37093-0010EA
86
USD
TS37093-0010
TS37093-0050
TS37093-0250
Ammonium hydrogen carbonate 98%, pure
Ammonium hydrogen carbonate
Formula:
NH₄HCO₃ MW: 79.06 g/mol Melting Pt: 35 (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012138 CAS Number: 1066-33-7 EINECS: 213-911-5 Merck Index: 13,00497 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration with HCl | ≥97.5 % |
Anticaking agent | ≤0.55 % (MgCO3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...