Lead(II) carbonate basic, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lead carbonat basic, White lead, Lead (II) carbonate hydroxide, Lead (II) hydroxide carbonate
AC22271-0025
AC22271-0050
AC22271-0100
AC22271-5000
TS22271-0025EA
220
USD
TS22271-0025
TS22271-0050
TS22271-0100
TS22271-5000
Lead(II) carbonate basic, extra pure
Lead(II) carbonate basic
Formula:
(PbCO₃)₂·Pb(OH)₂ Melting Pt: 400 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00078155 CAS Number: 1319-46-6 EINECS: 215-290-6 UN: 2291 ADR: 6.1,III |
Specification Test Results
Appearance | White to off-white Powder |
Titration Complexometric | ≥75 % Pb |
Insoluble matter | ≤0.05 % (in dilute acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...