Ammonium carbonate ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC40113-0010
AC40113-0025
AC40113-0250
TS40113-0010EA
150
USD
TS40113-0010
TS40113-0025
TS40113-0250
Ammonium carbonate ACS
Ammonium carbonate
Formula:
(NH₄)₂CO₃ Melting Pt: 58 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00010890 CAS Number: 506-87-6 EINECS: 208-058-0 Merck Index: 13,00508 |
Specification Test Results
Appearance | White Powder or crystals and/or chunks |
Non-volatile matter | ≤0.01 % |
Titration excess HCl | ≥30.0 % (NH3) |
Chloride (Cl) | ≤5 ppm |
Iron (Fe) | ≤5 ppm |
Heavy metals (ICP-OES) | ≤5 ppm |
Sulfur compounds (as SO4) | ≤0.002 % |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...