Ammonium carbonate, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Salt of hartshorn

AC39029-0010 AC39029-0025
TS39029-0010EA 89 USD
TS39029-0010 TS39029-0025
Ammonium carbonate, Extra Pure
Ammonium carbonate
Formula: (NH₄)₂CO₃
MW: 96.09 g/mol
Melting Pt: 58 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508

Order Now

Specification Test Results

Appearance White Powder or crystals and/or chunks
Non-volatile matter ≤100 ppm
Titration with HCl ≥30.5 % (NH3)
Chloride (Cl) ≤30 ppm
Sulfate (SO4) ≤30 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR