Lead(II) carbonate ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lead carbonat
AC21251-1000
AC21251-5000
TS21251-1000EA
82
USD
TS21251-1000
TS21251-5000
Lead(II) carbonate ACS
Lead(II) carbonate
Formula:
PbCO₃ Melting Pt: 400 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064631 CAS Number: 598-63-0 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | White Powder |
Chloride (Cl) | ≤0.002 % |
Nitrate and Nitrite (as NO3) | (limit about 0.005%) Passes test |
Cadmium (Cd) | ≤0.002 % |
Calcium (Ca) | ≤0.01 % |
Iron (Fe) | ≤0.005 % |
Potassium (K) | ≤0.02 % |
Sodium (Na) | ≤0.05 % |
Zinc (Zn) | ≤0.003 % |
Insoluble matter | ≤0.02 % (in dilute acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...