Barium perchlorate, anhydrous 99%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

AC22127-1000 AC22127-5000
TS22127-1000EA 55 USD
TS22127-1000 TS22127-5000
Barium perchlorate, anhydrous 99%, Extra Pure
Barium perchlorate
Formula: Ba(ClO₄)₂
Melting Pt: 505 °C
Density: 3.2 g/cm³
Flash Pt: (closed cup)
MDL Number: MFCD00003440
CAS Number: 13465-95-7
EINECS: 236-710-4
UN: 1447
ADR: 5.1,II
Merck Index: 12,01016

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric ≥98.5 %
Chloride (Cl) ≤1000 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤10 ppm
Nickel (Ni) ≤10 ppm
Insoluble matter ≤0.1 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR