Cobalt(II) chloride hexahydrate, pure

Supplier: Thermo Scientific Chemicals
Danger

AC19286-0010 AC19286-2500
TS19286-0010EA 267 USD
TS19286-0010 TS19286-2500
Cobalt(II) chloride hexahydrate, pure
Cobalt(II) chloride hexahydrate
Formula: CoCl₂·6H₂O
MW: 237.93 g/mol
Melting Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 13,02462

Order Now

Specification Test Results

Appearance Red to violet Crystals
Titration Complexometric 98.0 to 102.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR