Iodine monochloride ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Iodochloride

AC41236-1000 AC41236-5000
TS41236-1000EA 134 USD
TS41236-1000 TS41236-5000
Iodine monochloride ACS
Iodine monochloride
Formula: ClI
MW: 162.36 g/mol
Boiling Pt: 97.3
Melting Pt: 26
Density: 3.2 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00011354
CAS Number: 7790-99-0
EINECS: 232-236-7
UN: 1792
ADR: 8 II
Merck Index: 13,05039

Order Now

Specification Test Results

Appearance Dark red-brown to brown to black Liquid or solid
Assay 1.00 to 1.20 (I/Cl ratio of the Wijs solution prepared from the reagent)
Insoluble matter ≤0.005 %
GC ≥99.85 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR