Iodine monochloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Iodochloride

AC19696-1000 AC19696-5000
TS19696-1000EA 137 USD
TS19696-1000 TS19696-5000
Iodine monochloride 98%
Iodine monochloride
Formula: ClI
MW: 162.36 g/mol
Boiling Pt: 97.3
Melting Pt: 26
Density: 3.2 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00011354
CAS Number: 7790-99-0
EINECS: 232-236-7
UN: 1792
ADR: 8 II
Merck Index: 13,05039

Order Now

Specification Test Results

Appearance Dark red-brown to brown to black Liquid or solid
Titration Iodometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR