Trichloro(phenyl)silane 95%

Supplier: Thermo Scientific Chemicals
Danger

AC13100-0010 AC13100-0100 AC13100-2500
TS13100-0010EA 249 USD
TS13100-0010 TS13100-0100 TS13100-2500
Trichloro(phenyl)silane 95%
Trichloro(phenyl)silane
Formula: C₆H₅Cl₃Si
Boiling Pt: 201 °C (760 mmHg)
Melting Pt: –40 °C
Density: 1.321 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00000479
CAS Number: 98-13-5
EINECS: 202-640-8
UN: 1804
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5220 to 1.5240 (20°C, 589 nm)
GC ≥94 %
Hydroxyl value 65 to 78 mg KOH/g
Iodine value 25 to 35 g I2/100 g
Saponification value 65 to 70 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR